Learn More
Biomatik Corporation Bovine Lactoperoxidase (Lpo) Elisa Kit
Supplier:  Biomatik Corporation EKC31713
Encompass_Preferred
Bovine Lactoperoxidase (Lpo) Elisa Kit for detecting Lactoperoxidase in breast milk. Species Reactivity: Bovine (Bos taurus). Detection Range: 1.56-100mU/ml. Sensitivity: 1.56mU/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P80025; LPO. Target Synonyms: LPO; Lactoperoxidase; LPO; EC 1.11.1.7. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3644