Learn More
Biomatik Corporation Human Carbamoyl-Phosphate Synthase [Ammonia], Mitochondrial (Cps1) Elisa Kit
Supplier:  Biomatik Corporation EKC32977
Encompass_Preferred
Human Carbamoyl-Phosphate Synthase [Ammonia], Mitochondrial (Cps1) Elisa Kit for detecting Carbamoyl-Phosphate Synthetase 1, Mitochondrial in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5000pg/ml. Sensitivity: 19.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P31327; CPS1. Target Synonyms: Carbamoyl phosphate synthase [ammonia]; Carbamoyl phosphate synthase [ammonia] mitochondrial; Carbamoyl phosphate synthase; Carbamoyl phosphate synthetase 1; Carbamoyl phosphate synthetase 1 mitochondrial; Carbamoyl phosphate synthetase I; Carbamoyl-phosphate synthase [ammonia]; Carbamoyl-phosphate synthetase I; Carbamoylphosphate synthase; Carbamoylphosphate synthetase 1; Carbamoylphosphate synthetase I; CPS 1; Cps1; CPSase 1; CPSase I; CPSASE1; CPSM_HUMAN; mitochondrial; MS738. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3839