Learn More
Biomatik Corporation Human Inter-Alpha-Trypsin Inhibitor Heavy Chain H2 (Itih2) Elisa Kit
Supplier:  Biomatik Corporation EKC34181
Encompass_Preferred
Human Inter-Alpha-Trypsin Inhibitor Heavy Chain H2 (Itih2) Elisa Kit for detecting Inter-Alpha (Globulin) Inhibitor H2 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ug/ml. Sensitivity: 0.78ug/mL. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P19823; ITIH2. Target Synonyms: AI747202; H2P; IGHEP2; Inter alpha (globulin) inhibitor H2; Inter alpha globulin inhibitor H2 polypeptide; Inter alpha trypsin inhibitor complex component II; Inter alpha trypsin inhibitor heavy chain H2; Inter-alpha-inhibitor heavy chain 2; Inter-alpha-trypsin inhibitor complex component II; Inter-alpha-trypsin inhibitor heavy chain H2; Intin2; ITI HC2; ITI heavy chain H2; ITI-HC2; Itih 2; Itih2; ITIH2_HUMAN; OTTHUMP00000019058; OTTHUMP00000019059; OTTMUSP00000012019; RP23 119N4.3; Serum derived hyaluronan associated protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4058