Learn More
Biomatik Corporation Dog C Reactive Protein (CRP) ELISA Kit
Supplier:  Biomatik Corporation EKU10691
Encompass_Preferred
Dog C Reactive Protein (CRP) ELISA Kit for detecting CRP in serum, plasma, other biological fluids. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 0.156-10ng/ml. Sensitivity: 0.064ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: T2KEN6. Gene ID: 488629. Target Synonyms: C-RP; PTX1; Pentraxin-Related; Pentraxin 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3205