Learn More
Biomatik Corporation Human Heparin Cofactor II (HCII) Anti-antigen ELISA Kit
Supplier:  Biomatik Corporation EKU12520
Encompass_Preferred
Human Heparin Cofactor II (HCII) Anti-antigen ELISA Kit for detecting HCII in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.22ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h, 30min. Accession Number: P05546. Gene ID: 3053. Target Synonyms: SERPIND1; HC2; HCF2; HC II; HLS2; LS2; Serpin D1; Serpin Peptidase Inhibitor Clade D Member 1; Protease inhibitor leuserpin-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3542