Learn More
Biomatik Corporation Dog N-terminal Pro atrial natriuretic peptide (NT-ProANP)ELISA kit
Supplier:  Biomatik Corporation EKC32097
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Canis lupus familiaris (Dog) (Canis familiaris). Detection Range: 0.312 ng/mL-20 ng/mL. Sensitivity: 0.078 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-3704