Learn More
Biomatik Corporation Dog P-Selectin (Selp) Elisa Kit
Supplier:  Biomatik Corporation EKC32110
Encompass_Preferred
Dog P-Selectin (Selp) Elisa Kit for detecting Selectin P (Granule Membrane Protein 140Kda, Antigen Cd62) in serum, plasma, tissue homogenates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 12.5-800ng/ml. Sensitivity: 3.12ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Target Synonyms: RP1-86F14.2; CD62; CD62P; FLJ45155; GMP140; GRMP; LECAM3; PADGEM; PSEL; OTTHUMP00000032609; granulocyte membrane protein; leukocyte-endothelial cell adhesion molecule 3; platelet alpha-granule membrane. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3706