Learn More
Biomatik Corporation Human Beta-Hexosaminidase Subunit Alpha (Hexa) Elisa Kit
Supplier:  Biomatik Corporation EKC32866
Encompass_Preferred
Human Beta-Hexosaminidase Subunit Alpha (Hexa) Elisa Kit for detecting Hexosaminidase A (Alpha Polypeptide) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P06865; HEXA. Target Synonyms: Beta hexosaminidase alpha chain precursor; Beta hexosaminidase subunit alpha; Beta N acetylhexosaminidase; Beta N acetylhexosaminidase subunit alpha; Beta-hexosaminidase A; Beta-hexosaminidase subunit alpha; Beta-N-acetylhexosaminidase subunit alpha; Hexa; HEXA_HUMAN; Hexosaminidase A (alpha polypeptide); Hexosaminidase A alpha polypeptide; Hexosaminidase A; Hexosaminidase subunit A; MGC99608; N acetyl beta glucosaminidase; N acetyl beta glucosaminidase subunit alpha; N-acetyl-beta-glucosaminidase subunit alpha; TSD. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3827