Learn More
Biomatik Corporation Human cardiac troponin T(cTn-T)ELISA Kit
Supplier:  Biomatik Corporation EKC33014
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156 ng/mL-10 ng/mL. Sensitivity: 0.039 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P13805. Target Synonyms: ANM, FLJ98147, MGC104241, STNT, TNT, TNTS, slow skeletal muscle troponin T|troponin T1, skeletal, slow|troponin-T1, skeletal, slow. Restriction: For Research Use Only.
Catalog No. 50-229-3847