Learn More
Biomatik Corporation Human Furin (Furin) Elisa Kit
Supplier:  Biomatik Corporation EKC33765
Encompass_Preferred
Human Furin (Furin) Elisa Kit for detecting Furin (Paired Basic Amino Acid Cleaving Enzyme) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 23.5-1500pg/ml. Sensitivity: 5.86pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P09958; FURIN. Target Synonyms: FURIN; FUR; PACE; PCSK3; Furin; Dibasic-processing enzyme; Paired basic amino acid residue-cleaving enzyme; PACE. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3980