Learn More
Biomatik Corporation Human Glucose Transporter 4, Glut4 Elisa Kit
Supplier:  Biomatik Corporation EKC33830
Encompass_Preferred
Human Glucose Transporter 4, Glut4 Elisa Kit for detecting Solute Carrier Family 2 (Facilitated Glucose Transporter), Member 4 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P14672; SLC2A4. Target Synonyms: insulin-responsive; Glucose transporter GLUT 4; Glucose transporter type 4; Glucose transporter type 4 insulin responsive; GLUT 4; GLUT-4; GLUT4; GTR4_HUMAN; Insulin responsive glucose transporter type 4; kug; SLC 2A4; SLC2A4; solute carrier family 2 (facilitated glucose transporter) member 4; Solute carrier family 2 member 4; Solute carrier family 2; facilitated glucose transporter member 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3991