Learn More
Biomatik Corporation Human Histone H3.1 (Hist1H3A) Elisa Kit
Supplier:  Biomatik Corporation EKC34049
Encompass_Preferred
Human Histone H3.1 (Hist1H3A) Elisa Kit for detecting Histone Cluster 1, H3A in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 23.5-1500pg/ml. Sensitivity: 5.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P68431; HIST1H3A. Target Synonyms: H3 histone family member E pseudogene; H3 histone family; member A; H3/A; H31_HUMAN; H3F3; H3FA; Hist1h3a; HIST1H3B; HIST1H3C; HIST1H3D; HIST1H3E; HIST1H3F; HIST1H3G; HIST1H3H; HIST1H3I; HIST1H3J; HIST3H3; histone 1; H3a; Histone cluster 1; H3a; Histone H3 3 pseudogene; Histone H3.1; Histone H3/a; Histone H3/b; Histone H3/c; Histone H3/d; Histone H3/f; Histone H3/h; Histone H3/i; Histone H3/j; Histone H3/k; Histone H3/l. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4034