Learn More
Biomatik Corporation Rat Advanced glycosylation end product-specific receptor(AGER) ELISA kit
Supplier:  Biomatik Corporation EKC42368
Encompass_Preferred
Sample Type: serum, plasma,tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 31.25 pg/mL-2000 pg/mL. Sensitivity: 7.81 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q63495. Target Synonyms: DAMA-358M23.4, MGC22357, RAGE, receptor for advanced glycosylation end-products. Restriction: For Research Use Only.
Catalog No. 50-229-5063