Learn More
Biomatik Corporation Human AZU1 (Azurocidin) ELISA Kit
Supplier:  Biomatik Corporation EKF58190
Encompass_Preferred
Human AZU1 (Azurocidin) ELISA Kit for detecting AZU1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P20160. Target Synonyms: AZU1; Azurocidin 1; AZAMP; AZU1; CAP37; HBP; AZAMP; AZU; azurocidin; azurocidin 1; Azurocidin; Cationic Antimicrobial Protein-37; Heparin Binding Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5607