Learn More
Biomatik Corporation Mouse TFPI2 (Tissue Factor Pathway Inhibitor 2) ELISA Kit
Supplier:  Biomatik Corporation EKF58517
Encompass_Preferred
Mouse TFPI2 (Tissue Factor Pathway Inhibitor 2) ELISA Kit for detecting TFPI2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O35536. Target Synonyms: TFPI2; PP5; REF-1; Placental protein 5; PP5FLJ21164; TFPI-2REF1; tissue factor pathway inhibitor 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5833