Learn More
Biomatik Corporation Mouse NSE (Neuron-Specific Enolase) ELISA Kit
Supplier:  Biomatik Corporation EKF58591
Encompass_Preferred
Mouse NSE (Neuron-Specific Enolase) ELISA Kit for detecting NSE in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P17183. Target Synonyms: NSE; ENO2; gamma-Enolase; enolase 2; gamma; neuronal; gamma-enolase; Neural enolase; neuron specific gamma enolase; Neuron-specific enolase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5892