Learn More
Biomatik Corporation Human eosinophil cationic protein, ECP ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC33579
Encompass_Preferred
The Human eosinophil cationic protein, ECP ELISA Kit allows for quantitative determination of eosinophil cationic protein, ECP in serum, plasma, tissue homogenates. Species Reactivity: Human. Detection Range: 1.56 ng/mL-100 ng/mL. Sensitivity: 0.39 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. Restriction: For Research Use Only.
Catalog No. 50-149-7871