Learn More
Biomatik Corporation Human Myelin Protein P0 (Mpz) Elisa Kit
Supplier:  Biomatik Corporation EKC34691
Encompass_Preferred
Human Myelin Protein P0 (Mpz) Elisa Kit for detecting Myelin Protein Zero in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P25189; MPZ. Target Synonyms: Charcot Marie Tooth neuropathy 1B; CHM; CMT1; CMT1B; CMT2I; CMT2J; CMT4E; CMTDI3; CMTDID; DSS; HMSNIB; MPP; MPZ; Myelin peripheral protein; Myelin protein P0; Myelin protein zero; MYP0_HUMAN; P0. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4151