Learn More
Biomatik Corporation Mouse Β-Catenin, Β-Cat Elisa Kit
Supplier:  Biomatik Corporation EKC38071
Encompass_Preferred
Mouse Β-Catenin, Β-Cat Elisa Kit for detecting Catenin (Cadherin-Associated Protein), Beta 1, 88Kda in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q02248; Ctnnb1. Target Synonyms: Ctnnb1; CatnbCatenin beta-1; Beta-catenin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4488