Learn More
Biomatik Corporation Human Muscle Skeletal Receptor Tyrosine Kinase (Musk) Elisa Kit
Supplier:  Biomatik Corporation EKC34681
Encompass_Preferred
Human Muscle Skeletal Receptor Tyrosine Kinase (Musk) Elisa Kit for detecting Muscle, Skeletal, Receptor Tyrosine Kinase in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100pg/ml. Sensitivity: 0.39pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O15146; MUSK. Target Synonyms: MUSK; Muscle; skeletal receptor tyrosine-protein kinase; Muscle-specific tyrosine-protein kinase receptor; MuSK; Muscle-specific kinase receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4150