Learn More
Biomatik Corporation Human Occludin (Ocln) Elisa Kit
Supplier:  Biomatik Corporation EKC34871
Encompass_Preferred
Human Occludin (Ocln) Elisa Kit for detecting Occludin in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 23.5-1500pg/ml. Sensitivity: 5.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q16625; OCLN. Target Synonyms: OCLN; Occludin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4187