Learn More
Biomatik Corporation Mouse Aminopeptidase N(ANPEP) ELISA kit
Supplier:  Biomatik Corporation EKC36296
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 39 mU/mL-2500 mU/mL. Sensitivity: 9.7 mU/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P97449. Target Synonyms: APN, CD13, GP150, LAP1, P150, PEPN, aminopeptidase M|aminopeptidase N|membrane alanine aminopeptidase|microsomal aminopeptidase|myeloid plasma membrane glycoprotein CD13. Restriction: For Research Use Only.
Catalog No. 50-229-4363