Learn More
Biomatik Corporation Rat Cathepsin B (Ctsb) Elisa Kit
Supplier:  Biomatik Corporation EKC42454
Encompass_Preferred
Rat Cathepsin B (Ctsb) Elisa Kit for detecting Cathepsin B in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 18.75-1200ng/ml. Sensitivity: 4.67ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P00787; Ctsb. Target Synonyms: Ctsb; Cathepsin B; EC 3.4.22.1; Cathepsin B1; RSG-2) [Cleaved into: Cathepsin B light chain; Cathepsin B heavy chain]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5084