Learn More
Biomatik Corporation Human Nuclear Factor-Κb P65, Nf-Κb P65 Elisa Kit
Supplier:  Biomatik Corporation EKC40518
Encompass_Preferred
Human Nuclear Factor-Κb P65, Nf-Κb P65 Elisa Kit for detecting V-Rel Reticuloendotheliosis Viral Oncogene Homolog A (Avian) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 47-3000pg/ml. Sensitivity: 11.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q04206; RELA. Target Synonyms: Avian reticuloendotheliosis viral (v rel) oncogene homolog A; MGC131774; NF kappa B p65delta3; nfkappabp65; NFkB p65; NFKB3; Nuclear factor kappaB; Nuclear Factor NF Kappa B p65 Subunit; Nuclear factor NF-kappa-B p65 subunit; Nuclear factor of kappa light polypeptide gene enhancer in B cells 3; Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3; OTTHUMP00000233473; OTTHUMP00000233474; OTTHUMP00000233475. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4761