Learn More
Biomatik Corporation Rat Acan (Aggrecan core protein) ELISA Kit
Supplier:  Biomatik Corporation EKF58967
Encompass_Preferred
Rat Acan (Aggrecan core protein) ELISA Kit for detecting Acan in cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P07897. Target Synonyms: ACAN; Aggrecan core protein; Cartilage-specific proteoglycan core protein; CSPCP; Chondroitin sulfate proteoglycan core protein 1; Chondroitin sulfate proteoglycan 1; AGC1; CSPG1; MSK16; AGCAN; CSPGCP; SEDK. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6191