Learn More
Biomatik Corporation Human MLKL (mixed lineage kinase domain-like) ELISA Kit
Supplier:  Biomatik Corporation EKF59339
Encompass_Preferred
Human MLKL (mixed lineage kinase domain-like) ELISA Kit for detecting MLKL in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q8NB16. Target Synonyms: BC028141; MLKL. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6407