Learn More
Biomatik Corporation Mouse Ki67 (ProlifeRation marker protein Ki-67) ELISA Kit
Supplier:  Biomatik Corporation EKF59568
Encompass_Preferred
Mouse Ki67 (ProlifeRation marker protein Ki-67) ELISA Kit for detecting Ki67 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: E9PVX6. Target Synonyms: Ki67. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6415