Learn More
Biomatik Corporation EFNB3 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50192
Encompass_Preferred
EFNB3 Colorimetric Cell-Based ELISA Kit for detecting EFNB3 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q15768. Gene ID: 1949. Target Synonyms: EFL6; EPLG8; LERK8; ephrin-B3; Ephrin B3; LIGAND OF EPH-RELATED KINASE 8; eph-related receptor tyrosine kinase ligand 8; ephrin-B3 precursor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6914