Learn More
Biomatik Corporation Mouse NF-κB p65 (Nuclear Factor-κB p65) ELISA Kit
Supplier:  Biomatik Corporation EKF58276
Encompass_Preferred
Mouse NF-κB p65 (Nuclear Factor-κB p65) ELISA Kit for detecting NF-κB p65 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q04207. Target Synonyms: NF-κB p65; NFkB p65; NFKB3; p65RelA; MGC131774; NFKB3v-rel avian reticuloendotheliosis viral oncogene homolog A; nuclear factor ofkappa light polypeptide gene enhancer in B-cells 3; p65; Nuclear factor NF-kappa-B p65 subunit; Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3transcription factor p65; p65; v-rel reticuloendotheliosis viral oncogene homolog A; avian; v-rel reticuloendotheliosis viral oncogene homolog A; nuclear factor of kappalight polypeptide gene enhancer in B-cells 3; p65. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5669