Learn More
Biomatik Corporation Mouse IL-2sRα/CD25 (Soluble Interleukin-2 Receptor Alpha Chain) ELISA Kit
Supplier:  Biomatik Corporation EKF58412
Encompass_Preferred
Mouse IL-2sRα/CD25 (Soluble Interleukin-2 Receptor Alpha Chain) ELISA Kit for detecting IL-2sRα; CD25 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P01590. Target Synonyms: IL-2sRα; CD25; IL-2 R alpha; IL2RA; CD25; CD25 antigen; IDDM10; IL-2 receptor subunit alpha; IL2R; IL-2R subunit alpha; IL-2-RA; IL2-RA; interleukin 2 receptor; alpha; interleukin-2 receptor subunit alpha; p55; TAC antigen; TCGFR. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5760