Learn More
Biomatik Corporation Human S100A10 (Protein S100-A10) ELISA Kit
Supplier:  Biomatik Corporation EKF57508
Encompass_Preferred
Human S100A10 (Protein S100-A10) ELISA Kit for detecting S100A10 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P60903. Target Synonyms: S100A10; annexin II ligand; calpactin I; light polypeptide; ANX2L; ANX2LG; CAL1LGP11; 42C; Calpactin I light chain; Calpactin-1 light chain; Cellular ligand of annexin II; CLP11Ca[1]; MGC111133; p10; p10 protein; p11; P11; protein S100-A10; S100 calcium binding protein A10; S100 calcium binding protein A10; annexin II ligand; calpactin I; lightpolypeptide; p11; S100 calcium-binding protein A10; S100 calcium-binding protein A10; annexin II ligand; calpactin I; lightpolypeptide; p11. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5382