Learn More
Biomatik Corporation Human CD25 (Interleukin 2 Receptor Alpha) ELISA Kit
Supplier:  Biomatik Corporation EKF58178
Encompass_Preferred
Human CD25 (Interleukin 2 Receptor Alpha) ELISA Kit for detecting CD25 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 93.75-6000pg/ml. Sensitivity: 56.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P01589. Target Synonyms: CD25; IL-2Rα; Interleukin 2 Receptor Alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5598