Learn More
Biomatik Corporation Human GAP43 (Neuromodulin) ELISA Kit
Supplier:  Biomatik Corporation EKF58389
Encompass_Preferred
Human GAP43 (Neuromodulin) ELISA Kit for detecting GAP43 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P17677. Target Synonyms: GAP43; Growth Associated Protein 43; B-50; Basp2; PP46; Neuromodulin; Axonal membrane protein GAP-43; calmodulin-binding protein P-57; nerve growth-related peptide GAP43; Neural phosphoprotein B-50; neuron growth-associated protein 43; Growth-associated protein 43. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5746