Learn More
Biomatik Corporation Human ITIH1 (Inter-alpha-trypsin inhibitor heavy chain H1) ELISA Kit
Supplier:  Biomatik Corporation EKF58716
Encompass_Preferred
Human ITIH1 (Inter-alpha-trypsin inhibitor heavy chain H1) ELISA Kit for detecting ITIH1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.781-50ng/ml. Sensitivity: 0.469ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P19827. Target Synonyms: ITIH1; Inter-alpha-trypsin inhibitor heavy chain H1; ITI heavy chain H1; ITI-HC1; Inter-alpha-inhibitor heavy chain 1; Inter-alpha-trypsin inhibitor complex component III; Serum-derived hyaluronan-associated protein; SHAP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5990