Learn More
Biomatik Corporation Human S100A9 (Protein S100-A9) ELISA Kit
Supplier:  Biomatik Corporation EKF58955
Encompass_Preferred
Human S100A9 (Protein S100-A9) ELISA Kit for detecting S100A9 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.781-50ng/ml. Sensitivity: 0.469ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P06702. Target Synonyms: S100A9; S100 Calcium Binding Protein A9; Calgranulin B; MRP-14; CAGBMigRation inhibitory factor-related protein 14; Calgranulin-B; Calprotectin L1H subunit; CGLB; L1AG; LIAG; MRP14Leukocyte L1 complex heavy chain; NIF; P14; protein S100-A9. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6182