Learn More
Biomatik Corporation Human COL6 (Collagen Type VI) ELISA Kit
Supplier:  Biomatik Corporation EKF59012
Encompass_Preferred
Human COL6 (Collagen Type VI) ELISA Kit for detecting COL6 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.125-200ng/ml. Sensitivity: 1.875ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P12109; P12110. Target Synonyms: COL6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6229