Learn More
Biomatik Corporation COVID-19 Spike Protein ELISA Kit
Supplier:  Biomatik Corporation EKF60694
Encompass_Preferred
COVID-19 Spike Protein ELISA Kit for detecting 2019-nCoV(S) in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Coronavirus (COVID-19, SARS-CoV-2, 2019-nCoV). Detection Range: 0.78-50pg/ml. Sensitivity: 390fg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P0DTC2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6706