Learn More
Biomatik Corporation Human CA-125 (CarbohydRate Antigen 125) ELISA Kit
Supplier:  Biomatik Corporation EKF59184
Encompass_Preferred
Human CA-125 (CarbohydRate Antigen 125) ELISA Kit for detecting CA-125 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.563-100iu/ml. Sensitivity: 0.938iu/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q8WXI7. Target Synonyms: CA-125; CarbohydRate Antigen 125; CA125; MUC16; Mucin 16. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6371