Learn More
Biomatik Corporation ELISA KIT 96T PER KT
Supplier:  Biomatik Corporation EKU12707
Encompass_Preferred
Mouse Amyloid Precursor Protein (App) Mini Sample ELISA Kit for detecting APP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P12023. Gene ID: 11820. Target Synonyms: AAA; ABeta; ABPP; AD1; APPI; CVAP; PN2; CTFgamma; Peptidase Nexin-II Alzheimer Disease; Peptidase Nexin-II; Amyloid Beta (A4) Precursor Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-257-1266