Learn More
Biomatik Corporation Human soluble amyloid precursor protein (sAPP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35588
Encompass_Preferred
Human Soluble Amyloid Precursor Protein Α (Sappα) Elisa Kit for detecting Soluble Amyloid Precursor Protein Α (Sappα) in serum, plasma, tissue homogenates, cell culture supernates, cerebrospinal fluid (csf). Species Reactivity: Human (Homo sapiens). Detection Range: 12.5-800ng/ml. Sensitivity: 3.12ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Target Synonyms: soluble amyloid precursor protein alpha; sAPP alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8272