Learn More
Biomatik Corporation Human Sterol regulatory element-binding protein 1 (SREBF1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35669
Encompass_Preferred
Human Sterol Regulatory Element-Binding Protein 1 (Srebf1) Elisa Kit for detecting Sterol Regulatory Element Binding Transcription Factor 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 7.8-500pg/ml. Sensitivity: 1.95pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P36956; SREBF1. Target Synonyms: SREBF1; BHLHD1; SREBP1; Sterol regulatory element-binding protein 1; SREBP-1; Class D basic helix-loop-helix protein 1; bHLHd1; Sterol regulatory element-binding transcription factor 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8292