Learn More
Biomatik Corporation Rat MAOB ELISA Kit
Supplier:  Biomatik Corporation EKU12876
Encompass_Preferred
Rat MAOB ELISA Kit for detecting Monoamine Oxidase B in Cell Culture Supernates, Cell Lysates, Other Biological Fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.59ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P19643. Gene ID: 25750. Target Synonyms: MAO-B; Amine oxidase [flavin-containing] B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-000-15939