Learn More
Biomatik Corporation Human Vitamin D Receptor, VD R ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36035
Encompass_Preferred
Human Vitamin D Receptor, Vd R Elisa Kit for detecting Vitamin D (1, 25- Dihydroxyvitamin D3) Receptor in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 6.25-400pg/ml. Sensitivity: 1.56pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P11473; VDR. Target Synonyms: 1 25 dihydroxyvitamin D3 receptor; 1; 1; 25 dihydroxyvitamin D3 receptor; 1; 25-@dihydroxyvitamin D3 receptor; 25-dihydroxyvitamin D3 receptor; Member 1; NR1I1; Nuclear receptor subfamily 1 group I member 1; PPP1R163; Protein phosphatase 1; regulatory subunit 163; VDR; VDR_HUMAN; Vitamin D (1; 25- dihydroxyvitamin D3) receptor; Vitamin D hormone receptor; Vitamin D nuclear receptor variant 1; Vitamin D receptor; Vitamin D3 receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8356