Learn More
Biomatik Corporation Monkey N-terminal pro-brain natriuretic peptide, NT-proBNP ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36191
Encompass_Preferred
Monkey N-Terminal Pro-Brain Natriuretic Peptide, Nt-Probnp Elisa Kit for detecting N-Terminal Pro-Brain Natriuretic Peptide, Nt-Probnp in serum, plasma, tissue homogenates. Species Reactivity: Monkey (Simiiformes; infraorder). Detection Range: 0.78-50ng/ml. Sensitivity: 0.195ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8388