Learn More
Biomatik Corporation Mouse anti-nuclear Antibody (IgG) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36340
Encompass_Preferred
Mouse Anti-Nuclear Antibody (Igg) Elisa Kit for detecting Anti-Nuclear Antibody ( Igg) in serum, plasma. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.8-500pg/ml. Sensitivity: 1.95pg/ml. Format: 96-Well Plate. Assay Type: Capture ELISA. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8416