Learn More
Biomatik Corporation Mouse Complement C2 (C2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36643
Encompass_Preferred
Mouse Complement C2 (C2) Elisa Kit for detecting Complement Component 2 in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 18.75-300ng/ml. Sensitivity: 18.75ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P21180; C2. Target Synonyms: C2Complement C2; EC 3.4.21.43; C3/C5 convertase) [Cleaved into: Complement C2b fragment; Complement C2a fragment]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8452