Learn More
Biomatik Corporation Mouse complement fragment 3b (C3b) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36654
Encompass_Preferred
Mouse Complement Fragment 3B (C3B) Elisa Kit for detecting Complement Fragment 3B (C3B) in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.235-15ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8455