Learn More
Biomatik Corporation Mouse Interleukin 13, IL-13 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37150
Encompass_Preferred
Mouse Interleukin 13, Il-13 Elisa Kit for detecting Interleukin 13 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P20109; Il13. Target Synonyms: Il13; Il-13; Interleukin-13; IL-13; T-cell activation protein P600. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8555