Learn More
Biomatik Corporation Mouse L-lactate dehydrogenase A chain (LDH-A) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37299
Encompass_Preferred
Mouse L-Lactate Dehydrogenase A Chain (Ldh-A) Elisa Kit for detecting Lactate Dehydrogenase A in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10mU/ml. Sensitivity: 0.039mU/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P06151; Ldha. Target Synonyms: Ldha; Ldh-1; Ldh1L-lactate dehydrogenase A chain; LDH-A; EC 1.1.1.27; LDH muscle subunit; LDH-M. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8573