Learn More
Biomatik Corporation Mouse monocyte chemotactic protein 2, MCP-2 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37404
Encompass_Preferred
Mouse Monocyte Chemotactic Protein 2, Mcp-2 Elisa Kit for detecting Chemokine (C-C Motif) Ligand 8 in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 6.25-400pg/ml. Sensitivity: 1.56pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9Z121; Ccl8. Target Synonyms: Ccl8; Mcp2; Scya8C-C motif chemokine 8; Monocyte chemoattractant protein 2; Monocyte chemotactic protein 2; MCP-2; Small-inducible cytokine A8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8594